Learn More
Grant Instruments™ Universal Platform with Adjustable Bars
Adjust bars to fit a variety of vessels at once. Grant Instruments™ Universal Platform and Adjustable Bars does not require any tools to adjust bars.
Brand:  Grant Instruments™ PUP-12
1372.82 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10458633
  • 1410.00 DKK / Each
Only null left
Estimated Shipment: 25-09-2025
Description
  • For use with Grant Orbital Shakers and Shaker Incubators
  • Dimensions: 270 x 195mm
  • Includes Extra Holding Bars which can also be ordered separately (HB 200)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Universal Platform
285 x 220 mm
For Different Types Of Container
ES-20