Learn More
Graphite rod, pyrolytic coated, 2mm (0.08in) dia x 152mm (6in) long, Thermo Scientific Chemicals
1117.24 DKK - 4536.45 DKK
Specifications
Chemical Name or Material | Graphite rod |
---|---|
Name Note | Pyrolytic coated, 2mm (0.08 in.) dia. x 152mm (6 in.) long |
MDL Number | MFCD00144065 |
Solubility Information | It is insoluble in water. |
Physical Form | Rod |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15436545
|
Thermo Scientific Alfa Aesar
043670.KG |
1 Pc. |
1117.24 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15446545
|
Thermo Scientific Alfa Aesar
043670.KJ |
5 Pc. |
4536.45 DKK
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Graphite rod, is suitable for making: Anodes, Electrodes, Heating elements, Crucibles, Molds & Ingots, Hot press dies, Gas nozzles, Lubrication sticks, Bushings & bearings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Graphite rod | |
MFCD00144065 | |
Rod |
Pyrolytic coated, 2mm (0.08 in.) dia. x 152mm (6 in.) long | |
It is insoluble in water. |
Safety and Handling
missing translation for 'einecsNumber' : 231-955-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures