Learn More
Bollé Safety Visiteur Clear Safety Glasses
Shop All Bollé Safety Products
Click to view available options
Frame Material:
Polycarbonate
UV Protection:
Yes
For Use With (Application):
To protect eyes.
Unit Size:
Each
Description
- Polycarbonate
- Clear lenses
- Browguard
- Hinged sidearms
- Can be worn over spectacles
Specifications
Specifications
For Use With (Application) | To protect eyes. |
Frame Color | Clear |
Frame Material | Polycarbonate |
UV Protection | Yes |