Learn More
Mettler Toledo™ HA 405-DXK-S8 pH Combination Electrode
Used for a variety of tough chemical applications. Mettler Toledo™ HA 405-DXK-S8 pH Combination Electrode is a pressure-resistant, low-maintenance pH-electrode
Brand: Mettler Toledo™ 59903146
This item is not returnable.
View return policy
Description
- Low maintenance - A solid polymer electrolyte (no refilling and pressurizing of electrolyte) in direct contact with the process media via an open junction. No clogging in contaminated media due to the open junction.
- Long operational - electrode lifetime The electrode allows reliable process control in highly demanding chemical processes, particulary suitable for protein-containing solutions, emulsions and suspensions, hot caustic solutions, sulfide-bearing media etc..
- High pressure resistance - The electrode covers a generous pressure range and shows high resistance to process pressure fluctuations
Specifications
pH Combination Electrode | |
120 mm |