Learn More
Horst™ HBST Model Heating Tape
Brand:  Horst™ 20711
 View more versions of this product
Product Code. 10791913
  • 600.00€ / Each
Only null left
Estimated Shipment: 07-07-2025
Specifications
17 Lm
17 m
∼230V Voltage
820w
Heating Tape
17 Lm
230 V
HBST
25mm
250°C