Learn More
BRAND™ HDPE, Aspirator Bottle
Shop All Brand Products
Click to view available options
Capacity (Metric):
5 L
Unit Size:
Each
Description
- Hight Density Polyethylene (HDPE)
- Narrow neck
- Includes polyprpylene stopcock, screw cap and carrying handle
Specifications
Specifications
Capacity (English) | 1.3 gal. |
Capacity (Metric) | 5 L |
Closure Material | Polypropylene |
Diameter (Metric) Inner Neck | 45 mm |
Includes | Closure, Handles |
Material | HDPE |
Neck Style | Narrow |
Product Type | Aspirator Bottle |