Learn More
Heidolph™ Instruments Hei-VAP University Academic-Chill Package, EU-plug
The Hei-VAP University Academic-Chill Package features a hand lift, vacuum pump, chiller, and tubing set.
Brand:  Heidolph™ Instruments 571-25100-00
Product Code. 17363290
  • 75665.00 DKK / Each
Estimated Shipment: 24-06-2025
Description
  • Hei-VAP Core with hand lift and diagonal glassware set G1
  • Vacuum pump Rotavac Valve Tec
  • Manual vacuum controller
  • Chiller Hei-CHILL 250
  • Thermal fluid Kyro 30 (10 L)
  • Tubing set
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hand Lift Rotary Evaporator
EU-plug