Learn More
Fisherbrand™ Hexagonal Antistatic Weighing Boats
Shop All Fisherbrand Products
Click to view available options
Height (Metric):
9 mm
19 mm
22 mm
31 mm
Length (Metric):
38 mm
64 mm
102 mm
127 mm
Width (Metric):
25 mm
51 mm
76 mm
89 mm
Unit Size:
Pack of 500
Description
- Flexible boats can be bent at points to form pour spouts.
- Flat bottoms and sloping sides provide accurate transfer.
- Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
- Temperature range -10 to 70°C (14 to 158°F).
- Can be used with microwave moisture analyzers.
- Antistatic version only.
- 1 color, 4 sizes.
- Pack of 500.
- All are FDA/food-grade compliant.
- Disposable.
Specifications
Specifications
Bottom Shape | Flat |
Color | White |
Disposable | Single-use |
Material | Polystyrene |
Shape | Hexagonal |
Type | Weighing Boat |
For Use With (Equipment) | Microwave Moisture Analyzer |
Length (Metric) | 38 mm |
Width (Metric) | 25 mm |
Height (Metric) | 9 mm |