Learn More
Scientific Industries SI™ High-density Foam Inserts
Holds vessels in place while mixing. Scientific Industries SI™ High-density Foam Inserts fit snugly inside the Vortex-Genie 2 and Vortex-Genie Pulse Recessed Platform.
Brand:  Scientific Industries SI™ 503-0283-00
Product Code. 15537325
  • 409.00 DKK / Each
Only null left
Estimated Shipment: 18-09-2025
Description
  • For use with Vortex-Genie 2 and Vortex-Genie Pulse Recessed Platform
  • Accommodates:
    • 9-16mm Tubes
    • 14-29mm Tubes
    • 29-37mm Tubes
  • Blank Foam Insert provides customization for oddly-shaped vessels
  • All items included in Large Sample Set (Modle 302)
Specifications
Specifications
Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse for beakers, flasks and large tubes.
An assortment of Foam Inserts for tubes of various diameters (9mm to 37mm), 1 Blank Foam Insert for customization.
Recessed Platform 15547325