Learn More
Honeywell™ Freedom Half-Mask Air-Purifying Respirator (APR)
Click to view available options
Cartridge/Filter:
A1
A1B1E1K1P3
A1P1
A2
A2P3
P3
Unit Size:
Each
Description
- Product Type: Air-Purifying Respirator (APR)
- Mask Type: Half Mask
- Standards: EN405
- Cartridge Type: Dual Cartridge, Pre-Assembled
- Fastenings: One-Piece Sliding Suspension System
- Hazards: Gases, Organic Vapors (Temp. >65°C), Inorganic Vapors, Acids, Ammoniac and Derived Aminates (Class 1); Solid and Liquid Particulates (Class 3)
- For Use With (Equipment): Prefilter Pads (11847171, 12682743) and Pad Retaining Covers (12662653)
- For Use With (Applications): Chemical Industry, Construction Industry, Food Services, Manufacturing Industry, Medical Industry, Pharmaceutical Industry
Specifications
Specifications
Cartridge/Filter | A1B1E1K1P3 |
Certifications/Compliance | EC 394, CEN TC79 EN 405 |
Certifications (EN) | EC 394, CEN TC79, EN 405 |
Color | Green, Yellow, Gray, Brown |
Design Type | Cup/Molded |
Disposable | Reusable |
Product Type | Respiratory Protection |
For Use With (Application) | For protection against gas and organic vapors |
For Use With (Equipment) | Prefilter Pads [11847171 (P1), 12682743 (P2)] and Pad Retaining Covers (12662653) |
Quantity | 1 Ea. |