Learn More
Honeywell KCL™ Honeywell KCL™ Vitoject™ 890 Gloves
Made of Viton. Honeywell KCL™ Honeywell KCL™ Vitoject™ 890 Gloves have superb quality and are chemically resistant. The gloves are excellent for working with aggressive chemicals.
2435.00 DKK
Specifications
Certifications/Compliance | IFA 1301115 |
---|---|
Color | Black |
Latex Free | Yes |
Material | Viton |
Product Type | Gloves |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
10105374
|
Honeywell KCL™
089010111X |
10 |
2435.00 DKK
1 pair |
Estimated Shipment: 18-11-2025
Log in to see stock. |
|||||
10115374
|
Honeywell KCL™
089011111X |
11 |
2435.00 DKK
1 pair |
Estimated Shipment: 25-11-2025
Log in to see stock. |
|||||
10339264
|
Honeywell KCL™
089009111X |
9 |
2435.00 DKK
1 pair |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||||
Description
- AQL < 0.65
- Loose fit allows heat-protecting gloves to be worn underneath
- Can be re-used after contamination with certain chemicals, after special cleaning process (please consult KCL Laboratories)
- Can be combined with chemical protective clothing
- Excellent low degradation
- Good resistance against polycyclic aromatic hydrocarbons (petrochemicals
Specifications
IFA 1301115 | |
Yes | |
Gloves | |
340-360 cm |
Black | |
Viton | |
0.7 cm |