Learn More
Horst™ HSS Model Flexible Heating Conductor
Brand:  Horst™ 20903
104.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10097081
  • 339.00€ / Each
Only null left
Estimated Shipment: 31-07-2025
Specifications
2 Lm
∼230V Voltage
Flexible Heating Conductor
2 Lm
HSS