Learn More
HYDRANAL™ - Coulomat AG-H, Reagent for coulometric KF titration in long-chained hydrocarbons (anolyte solution), for cells with and without diaphragm, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
6 x 500 mL
Unit Size:
500mL
Pack of 6
Specifications
Specifications
Chemical Name or Material | HYDRANAL™-Coulomat AG-H |
Flash Point | 11°C (51.8°F) |
Quantity | 500 mL |
CAS | 67-56-1, 112-18-5, 71-41-0 |
UN Number | UN1992 |
Safety and Handling
Product Identifier
- HYDRANAL
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 3
- Reproductive toxicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 1 B
- H225-Highly flammable liquid and vapour.
- H311 + H331-Toxic in contact with skin or if inhaled.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.,Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.,Causes serious eye damage.
- H311-Toxic in contact with skin.
- H335-May cause respiratory irritation.,May cause respiratory irritation.
- H331-Toxic if inhaled.
- H360D-May damage the unborn child.,May damage the unborn child.
- H373-May cause damage to organs.,May cause damage to organs.
- H370-Causes damage to organs.,Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.,Toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : methanol, 1-pentanol, Dodecyldimethylamine, imidazole, sulphur dioxide, 1H-Imidazole monohydriodide