missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 37%, ACS Reagent Grade, Scharlab

Product Code. 11372465
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a laboratory reagent, acidifying agent, and in the production of chlorides and the synthesis of organic products.

  • Synonyms: Hydrochloric acid fuming, Muriatic acid, Hydrogen chloride solution
  • Molecular Formula: HCl
  • Molar Mass: 36.46g/mol
  • CAS: 7647-01-0
  • EIN ECS No.: 231-595-7
  • Residue After Evaporation: 0.001% max

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Appearance of Solution Passes Test
Melting Point -28°C
Boiling Point ∼50°C
Density ∼1.19g/cm3
Identification Passes Test
Residue after Ignition 0.0003% max
Vapor Pressure 190hPa at 20°C
Quantity 2.5 L
Physical Form Solution
Solubility Information Miscible in water (20°C)
Formula Weight 36.46g/mol
Percent Purity 36.5 to 38.0% (acidimetric)
Grade ACS Reagent
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid, 37%, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.