Learn More
Amarell™ Hygro-Thermometer
Provides fast readings with semi-conductor probe. The Hygro-Thermometer provides readings between 0 to 50°C and 20 to 90% rF.
Brand: Amarell™ E915020
This item is not returnable.
View return policy
Description
- Includes device for wall mounting (with ring or double-sided adhesive tape)
- Min./Max. function
- °C ↔ °F
- Fast responding semi-conductor probe
Specifications
Thermohygrometer | |
1% rF (rh) | |
0°C to 50°C | |
0.1°C | |
Probe |
20 to 90% | |
±5% rF (rh) | |
±1°C | |
65 x 97 x 20 mm |