Learn More
Ansell™ Hynit™ 32-105 Series Blue Nitrile Medium Weight Gloves
Special nitrile formulation unites excellent resistance to abrasion with remarkable flexibility. Liquid-repellent impregnation repels oil, grease and dirt.
362.00 DKK
Specifications
Back Material | Nitrile |
---|---|
Certifications/Compliance | CE Mark, ANSI, EN 388 : 3111 |
Cuff Material | Nitrile |
Coating Material | Nitrile |
Style | 32-105 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12680822
|
Ansell™
32105070 |
7 |
362.00 DKK
12 pairs |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
12690822
|
Ansell™
32105075 |
7.5 |
362.00 DKK
12 pairs |
Estimated Shipment: 12-08-2025
Log in to see stock. |
|||||
Description
Ideal for general plant use, assembly inspection, receiving, and light to medium fabrication and materials handling.
- Nitrile-impregnated fabric ensures excellent abrasion and snag resistance
- Feature clute cut and interlock knit liner for superior dry grip and excellent dexterity
- Gloves are liquid repellent and machine washable so they will not shrink, stiffen, or lose their grip
- Lining will not ball up
- Color-coded cuffs for easy size identification and size mating after laundering
- Available with solid and air-conditioned backs
Specifications
Nitrile | |
Nitrile | |
32-105 | |
Interlock Knit Polyester Cotton | |
144 Pairs | |
32-105 |
CE Mark, ANSI, EN 388 : 3111 | |
Nitrile | |
Slip-on | |
Fully coated-palm | |
Excellent snag, puncture, abrasion and cut resistance |