Learn More
Ambion™ mirVana™ miRNA Inhibitor
mirVana™ miRNA Inhibitor
Brand:  Ambion™ 4464085
8136.00 DKK valid until 2025-12-31
Use promo code "24071" to get your promotional price.
 View more versions of this product
Product Code. 13416706
  • 10170.00 DKK / 20nmol
Cell Biology Promotion
Use promo code 24071 at checkout
View Available Promotions
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store in freezer (-5 to -30°C).
mirVana™ miRNA Inhibitor, 20nmol, Small, Chemically Modified Single-Stranded RNA Molecules, Designed to Specifically Bind to and Inhibit Endogenous miRNA, Molecules and Enable miRNA Functional Analysis by Down-Regulation of miRNA Activity, For Research Use Only
mirVana™ miRNA Inhibitor
20 nmol