Learn More
Molecular Probes™ IANBD Amide (N,N'-Dimethyl-N-(Iodoacetyl)-N'-(7-Nitrobenz-2-Oxa-1,3-Diazol-4-yl)Ethylenediamine)
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
The thiol-reactive IANBD amide can be used to create environment sensitive probes. The absorption and fluorescence emission spectra, quantum yields, and extinction coefficients of NBD conjugates are all markedly dependent on solvent. Additionally, NBD is also a function analog of the dinitrophenyl hapten, and its fluorescence is quenched upon binding to anti-dinitrophenyl (anti-DNP) antibodies.
Specifications
Specifications
Quantity | 25 mg |
Product Type | IANBD Amide |
Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
Chemical Reactivity | Thiol |
Shipping Condition | Room Temperature |
Reactive Moiety | Alkyl Halide, Iodoacetamide |
Label or Dye | NBD |
Label Type | Classic Dyes |