Learn More
Manganese Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep CLMS-1
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ce | 10μg/mL |
Dy | 10μg/mL |
Er | 10μg/mL |
Eu | 10μg/mL |
Gd | 10μg/mL |
Ho | 10μg/mL |
La | 10μg/mL |
Lu | 10μg/mL |
Nd | 10μg/mL |
Pr | 10μg/mL |
Sc | 10μg/mL |
Sm | 10μg/mL |
Tb | 10μg/mL |
Th | 10μg/mL |
Tm | 10μg/mL |
Y | 10μg/mL |
Yb | 10μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Multi-Element Solution 1 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
ICP/ICP-MS Standard | |
125 mL | |
Each component at 10μg/mL in 5% HNO3:, Cerium, Dysprosium, Erbium, Europium, Gadolinium, Holmium, Lanthanum, Lutetium, Neodymium |
Triple-leached LDPE bottle | |
ICP-MS | |
Standard Solution | |
5% HNO3 | |
ICP/ICP-MS |
Product intended for laboratory use only