missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-19 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23705-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a cytokine that belongs to the IL10 cytokine subfamily. This cytokine is found to be preferentially expressed in monocytes. It can bind the IL20 receptor complex and lead to the activation of the signal transducer and activator of transcription 3 (STAT3). A similar cytokine in mouse is reported to up-regulate the expression of IL6 and TNF-alpha and induce apoptosis, which suggests a role of this cytokine in inflammatory responses. Alternatively spliced transcript variants encoding the distinct isoforms have been described.Specifications
IL-19 | |
Polyclonal | |
Unconjugated | |
IL19 | |
IL 10C, IL 19, IL19, interleukin 19, MDA1, NG.1, ZMDA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29949 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UHD0 | |
IL19 | |
IL-19 Fusion Protein Ag18887 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction