Learn More
IL-19 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23705-1-AP-20UL
 View more versions of this product
Product Code. 16862415
  • 1081.84 DKK / 20µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The protein encoded by this gene is a cytokine that belongs to the IL10 cytokine subfamily. This cytokine is found to be preferentially expressed in monocytes. It can bind the IL20 receptor complex and lead to the activation of the signal transducer and activator of transcription 3 (STAT3). A similar cytokine in mouse is reported to up-regulate the expression of IL6 and TNF-alpha and induce apoptosis, which suggests a role of this cytokine in inflammatory responses. Alternatively spliced transcript variants encoding the distinct isoforms have been described.
Specifications
Specifications
IL-19
Polyclonal
Unconjugated
IL19
IL 10C, IL 19, IL19, interleukin 19, MDA1, NG.1, ZMDA1
Rabbit
Antigen Affinity Chromatography
RUO
29949
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UHD0
IL19
IL-19 Fusion Protein Ag18887
20 μL
Primary
Human
Antibody
IgG