Learn More
GPE Scientific™ J Young Air Inlet Valve
GPE Scientific™ J Young Air Inlet Valve provides fine control for bleeding down a vacuum system.
Brand: GPE Scientific™ ALS/1
This item is not returnable.
View return policy
Description
- Many vacuum techniques require the system to be bled down to air.
- This valve fulfills this requirement simply by turning the screw cap which releases a PTFE piston.
- Fine control.
Specifications
Air Inlet Valve | |
Fine thread | |
5 mm bore |