Learn More
Fisherbrand™ Jet Spikes
Jet spikes are always delivered with nozzles.
586.00 DKK - 650.00 DKK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
17157914
|
Fisherbrand™
8300105 |
200 mm |
586.00 DKK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17167914
|
Fisherbrand™
8300106 |
270 mm |
618.00 DKK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
17177914
|
Fisherbrand™
8300107 |
360 mm |
650.00 DKK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
|||||
Description
- Equipped with nozzle
- For threaded units
Specifications
Jet Spikes | |
Nozzle |