Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Bonded White PTFE Liners
Intended for single-use injection
5740.00 DKK
Specifications
Material (Liner) | PTFE |
---|---|
Material | Polypropylene |
Autoclavable | Autoclavable |
Product Code | Brand | Color | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Closure Size | Price | Quantity & Availability | ||||
11833043
|
DWK Life Sciences
73814WH8425 |
White | 8-425 |
5740.00 DKK
Pack of 1000 |
Estimated Shipment: 03-11-2025
Log in to see stock. |
||||
11823043
|
DWK Life Sciences
73814BL9425 |
Blue | 9-425 |
5740.00 DKK
Pack of 1000 |
Estimated Shipment: 03-11-2025
Log in to see stock. |
||||
11813043
|
DWK Life Sciences
73814BK13425 |
Black | 13-425 |
5740.00 DKK
Pack of 1000 |
Estimated Shipment: 03-11-2025
Log in to see stock. |
||||
Description
- Manufactured using the highest quality raw materials
- Designed for an ideal fit with Kimble Chase autosampler vials
- Pre-assembled caps and septa minimizes contamination due to handling
Specifications
PTFE | |
Polypropylene | |
Autoclavable |