Learn More
Kova Plastics by Alltrista Glasstic™ Slide 10 with Grids
Description
Use optically clear plastic for microscopic analysis with same refractive index as glass. Glasstic™ Slide 10 with Grids is a 10-chambered slide with a hemocytometer-type grid.
Glasstic Slides sets the standard for accurate, reproducible, cost-effective and time-efficient standardized microscopic analysis. Glasstic Slides are extremely time-efficient and cost-effective.
- Manufactured from optically clear plastic with same refractive index as glass
- Each 10-chambered Glasstic Slide features 10 individual fully segregated specimen wells
- Capillary action reduces pipetting errors
- Suspends sediments homogeneously throughout the specimen
- Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
- Chamber walls prevent specimen leakage and cross-contamination
- Glasstic slide grid is in the focal plane of the specimen, allowing for use over a range of magification
- Provides results that have high correlation with results obtained with precision hemocytometers: red blood cells: r: 0.99; white blood cells: r: 0.99; platelets: r: 0.93
Specifications
Specifications
Type | Glasstic Slide 10:100 Slides with Grids |
For Use With (Application) | Microscopic examination of Urine and other body fluids |
Gridded | Yes |
Material | Plastic with the optical qualities of glass |
No. of Wells | 10 |
Quantity | Each |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.