Learn More
Labconco™ California Büchner Funnel
For use with Labconco Crude Fiber Apparatus.
Brand: Labconco™ 5510000
1434.29 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Stainless steel and polyethylene, two piece Buchner Funnel is modified by the addition of a 200-mesh stainless steel screen that is firmly heat sealed into the bottom of the upper section.Specifications
Polyethylene | |
Crude fiber apparatus | |
Buchner Funnel |