Learn More
Labconco™ CentriVap™ Complete Concentrator
Comes with everything needed to start processing
Brand: Labconco™ 7315030
128044.72 DKK valid until 2025-12-20
Use promo code "24121" to get your promotional price.
Description
Comes with everything needed to start processing
Uses a combination of centrifugal force, vacuum and heat to speed evaporation.
Includes:
- Vacuum Concentrator
- -50°C Cold Trap
- Aluminum rotor holds 40 microcentrifuge tubes and 12-13mm tubes
- Chemical-resistant diaphragm vacuum pump
- CentriZap Strobe Light allows user to view samples while rotor is spinning and lid is closed
Molecular biology, proteomics, genomics, cell biology, microbiology, biochemistry, drug discovery and analytical chemistry
Specifications
Centrivap | |
International | |
Heat Boost Not Included | |
230 V 50 Hz 6 A |