Learn More
Labconco™ Manual Valve Gas and Vacuum Tubing Connection Kit for Precise™ Glove Boxes
Accessory for Precise Glove Boxes
Brand: Labconco™ 5245100
This item is not returnable.
View return policy
Description
- Includes flexible vinyl tubing and fittings
- Connects user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
Specifications
Precise™ Basic™ Glove Box and Controlled Atmosphere Glove Box | |
20 lb. | |
9.1 Kg | |
Manual Valve Gas and Vacuum Tubing Connection Kit |