Learn More
Smeg™ Laboratory Glassware Washer
Clean a wide range of glassware with this washer. Smeg™ Laboratory Glassware Washers are manufactured with high quality materials and feature faultless, intuitive performance. Specific accessories ensure high quality washing of even the most difficult and dirtier instruments.
4375.00€ - 13625.00€
Specifications
Material | Stainless Steel |
---|---|
Product Type | Glassware Washer |
Dimensions (D x W x H) Interior | 490 x 525 x 570mm |
Product Code | Brand | Includes | Electrical Requirements | Temperature Setting | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Electrical Requirements | Temperature Setting | Price | Quantity & Availability | |||
15354962
|
Smeg™
GW0160S |
7 default programs | 3/N/PE 400V - 50Hz - 7 kW max | Washing up to 85°C | |||||
15375052
|
Smeg™
GW1160C |
9 default programs + 6 free | 3/N/PE 400V - 50Hz - 7 kW max | Washing up to 95°C | |||||
11932699
|
Smeg™
GW3060S |
20 default programs + 10 free | 3/N/PE 400V - 50Hz - 7 kW max (switchable single-phase 230V) | Washing up to 95°C |
9520.00€
Each |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||
15387333
|
Smeg™
GW4060S-1 |
9 default programs + 6 free | 1/N/PE 230V - 50Hz - 2,8 kW max | Washing up to 95°C | |||||
15374962
|
Smeg™
GW0160S1 |
7 default programs | 1/N/PE 230V - 50Hz - 2,8 kW max | Washing up to 85°C | |||||
11942699
|
Smeg™
GW4090 |
20 default programs + 10 free, active thermodynamic drying | 3/N/PE 400V - 50Hz - 7 kW max (switchable single-phase 230V) | Washing up to 95°C |
13625.00€
Each |
Estimated Shipment: 27-10-2025
Log in to see stock. |
|||
15364962
|
Smeg™
GW0160 |
7 default programs, alkaline powder detergent dispenser | 3/N/PE 400V - 50Hz - 7 kW max | Washing up to 85°C | |||||
Description
- Microprocessor electronic control
- Active thermodynamic drying
- Alkaline powder detergent dispenser
- Peristaltic pump for acid liquid neutralizer
- Washing chamber and internal door in AISI 304 stainless steel
- External panels made in AISI 304 stainless steel
- 2 independent washing levels through telescopic guides
- Incorporated water softener with salt sensor
- Cold water and demineralized water connections
- Electronic door lock system for user safety
- Without trolleys
Specifications
Stainless Steel | |
Glassware Washer | |
490 x 525 x 570mm |