Learn More
Uvex™ Ladies Coat 7463-Graphit
Well thought-out functionality, sporty design, exciting color contrasts, unusual material combinations and a cut that fits like a glove.
585.00 DKK
Specifications
Gender | Women's |
---|---|
Material | Cotton, Polyester |
Color | Gray |
Closure Type | Press Studs |
Disposable | Reusable |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17205711
|
Uvex™
8825909 |
Small |
585.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
17215711
|
Uvex™
8825910 |
Medium |
585.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
17225711
|
Uvex™
8825911 |
Large |
585.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
17235711
|
Uvex™
8825912 |
X-Large |
585.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
17245711
|
Uvex™
8825913 |
2X-Large |
585.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
17255711
|
Uvex™
8825914 |
3X-Large |
585.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
17265711
|
Uvex™
8825915 |
4X-Large |
585.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
17295701
|
Uvex™
8825908 |
X-Small |
585.00 DKK
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
|||||
Description
- Women coat
- Material: 66.5% polyester, 32% cotton, 1.5% antistatic. Fibers, 245 g / m²
- Color white
- Highest protection with antistat.
- Fiber - optimal charge reduction
- High rise arm construction
- Breast pocket on the left
- Two side pockets
- Turn-down collar standards: IEC 61340, OEKO-TEX™ Standard 100
- Sizes: XS-4XL
- Oversize surcharge: from size XXL + 10%
- Normal wash 60 ° C, do not bleach, dry Dryer, iron 2 points, chem.
Specifications
Women's | |
Gray | |
Reusable | |
245 gsm |
Cotton, Polyester | |
Press Studs | |
General Purpose Jacket |