Learn More
Brady™ Laminated Polyester: ISO Safety Sign - Warning; Crushing of hands
Sign compliant to ISO 7010.
30.85 DKK - 73.45 DKK
Specifications
Finish | Gloss |
---|---|
Material | Polyester |
Adhesive Type | Permanent Acrylic |
For Use With (Application) | Indoor and Outdoor |
Thickness | 0.099 mm |
Product Code | Brand | Width (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
15614757
|
Brady™
829869 |
50 mm | 43 mm |
46.95 DKK
Pack of 7 |
In Stock
Log in to see stock. |
||||
15624757
|
Brady™
829871 |
100 mm | 87 mm |
73.45 DKK
Pack of 3 |
In Stock
Log in to see stock. |
||||
15634757
|
Brady™
829872 |
200 mm | 173 mm |
30.85 DKK
Each |
In Stock
Log in to see stock. |
||||
15644757
|
Brady™
829873 |
315 mm | 273 mm |
52.40 DKK
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||
Description
- Laminated polyester
- For indoor and outdoor use
Specifications
Gloss | |
Permanent Acrylic | |
0.099 mm | |
Triangular | |
Black on Yellow |
Polyester | |
Indoor and Outdoor | |
-40°C to 110°C | |
Global |