Learn More
Nabertherm™ Lid for Square Saggars
Lid for square saggar for furnaces LHTC and LH.
Brand:  Nabertherm™ 699000985
35.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15350345
  • 35.90€ / Each
Only null left
Estimated Shipment: 08-09-2025
Description
  • Tmax: 1600°C
Specifications
Specifications
1600°C
Lid