Learn More
Lithium Tetraborate/Lithium Iodide (99.5%/0.50%) Fusion Flux Pure Crystal For Sample Preparation, SPEX CertiPrep™
Product Code. 15226056 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 kg
Packaging:
Triple-leached LDPE bottle
Unit Size:
1kg
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Lithium Tetraborate/Lithium Iodide (99.5%/0.50%), Pure Grade Fusion Flux for Sample Preparation, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
  • Pure Grade Fusion Flux
  • 1 kg
  • Lithium Tetraborate/Lithium Iodide (99.5%/0.50%)
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Quantity 1 kg
Grade Pure
Concentration or Composition (by Analyte or Components) Various
Packaging Triple-leached LDPE bottle
Chemical Name or Material Lithium Tetraborate/Lithium Iodide, Fusion Flux
Product intended for laboratory use only