Learn More
Lovibond™ PD250 Reagent Dispenser Refill: DPD Free Chlorine and Total Chlorine
For use with SC450 Chlorine and pH colorimeter, MC500 colorimeter and SP600 spectrophotometer
Brand: Lovibond™ 530160
693.87 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Specifications
For chlorine testing | |
Includes 1 vial of DPD free chlorine reagent and 1 vial of DPD total chlorine reagent | |
Replacement Free and Total Chlorine Reagent |
Used with SC450 chlorine and pH colorimeter, MC500 colorimeter and SP600 spectrophotometer | |
250 Tests |
Safety and Handling
- 2 x 250 TESTS VARIO POWDER FREE&TOTAL CHLORINE .
Signal Word
- Danger
Hazard Category
- Specific target organ toxicity Category 1
Hazard Statement
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P314-Get medical advice/attention if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : potassium iodide, EDTA