Learn More
Witeg™ Low Form Glass Beakers with Spout
With graduations for accurate and precise filling and dispensing.
23.05€ - 104.00€
Specifications
Color | Transparent |
---|---|
Material | Borosilicate Glass |
Includes | Spout |
Type | Beaker |
Graduated | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
13497537
|
Witeg™
5 500 150 S |
150 mL |
23.05€
Pack of 10 |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
13417547
|
Witeg™
5 500 250 S |
250 mL |
23.10€
Pack of 10 |
Estimated Shipment: 06-10-2025
Log in to see stock. |
|||||
13467577
|
Witeg™
5 500 400 |
400 mL | |||||||
13437557
|
Witeg™
5 500 600 S |
600 mL |
42.35€
Pack of 10 |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
Description
- Wide range of capacities available
- Acc. to ISO 3819
- Manufactured from DURAN™ borosilicate glass 3.3
- With spout for easy pouring
Specifications
Transparent | |
Spout | |
Yes | |
Low Form |
Borosilicate Glass | |
Beaker | |
ISO 3819 |