Learn More
Agilent BioTek Magnet Adapter Kit
GREENER_CHOICE
Magnet adapter kit for use with the 406 FX Washer Dispenser.
Brand:  Agilent BioTek 2120084
Product Code. 17804677
  • 1050600.00 DKK / Each
Description
  • Magnet sold separately
  • Enables magnetic bead washing and dispensing
  • BioStack compatible