Learn More
Thermo Scientific™ MassRuler DNA Ladders, ready-to-use
Thermo Scientific MassRuler DNA Ladders are designed for quantification and sizing of DNA fragments by agarose gel electrophoresis.
1046.00 DKK - 1214.00 DKK
Specifications
Concentration | 6X |
---|---|
Gel Type | Agarose |
Label or Dye | Bromophenol Blue |
Ready to Load | Yes |
Gel Compatibility | Agarose Gels |
Product Code | Brand | Size Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size Range | Price | Quantity & Availability | |||||
10849540
![]() |
Thermo Scientific™
SM0383 |
80 to 1031 bp |
1128.00 DKK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10170690
![]() |
Thermo Scientific™
SM0403 |
80 to 10000 bp |
1192.00 DKK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10736041
![]() |
Thermo Scientific™
SM1263 |
100 to 1000 bp |
1046.00 DKK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10746231
![]() |
Thermo Scientific™
SM1283 |
100 to 1000 bp |
1214.00 DKK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10374380
![]() |
Thermo Scientific™
SM0393 |
1500 to 10000 bp |
1094.00 DKK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific MassRuler DNA Ladders are designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis. Individuals DNA fragments are chromatography-purified and calibrated against a standard that guarantees precise quantity of each band.
Highlights
- Accurate DNA sizing and quantification – DNA ladder fragments are quantified spectrophotometrically
- Sharp bands, easy-to-remember fragment sizes and quantities
- Ready-to-use—premixed with 6X MassRuler DNA Loading Dye for direct loading
- Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use MassRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
Specifications
6X | |
Bromophenol Blue | |
Agarose Gels | |
MassRuler |
Agarose | |
Yes | |
Sustainable packaging | |
2 x 500 μL |
Safety and Handling
missing translation for 'shelfLife' : 24 Months when stored at room temperature.