Learn More
McFarland Standard 0.5, Pro-Lab Diagnostics™
Click to view available options
Quantity:
10 mL
Unit Size:
Each
Specifications
Specifications
Packaging | 10mL |
Physical Form | Liquid |
Color | Opaque |
Solubility | Soluble in water |
Quantity | 10 mL |