Learn More
AMPri™ Med-Comfort Premium Grip Latex Examination Gloves, PPE Cat. III
Non-sterile AMPri™ Med-Comfort latex examination gloves are powder-free and have a textured finish for good grip. Ideal for general laboratory use as well as clinical, pharmaceutical and food use applications, they offer excellent protection against microorganisms (viral, bacteria and fungi).
3.75 DKK - 286.98 DKK
Specifications
Certifications/Compliance | AQL 1.5 |
---|---|
Certifications/Compliance (Europe) | EN 455-1, EN374-1/4/5, EN420, Food grade |
Color | White |
Disposable | Single-use |
Laboratory/Cleanroom Use | Laboratory |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16826081
|
AMPri™
01037S |
Small |
286.98 DKK
Pack of 100 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||||
16816081
|
AMPri™
01037M |
Medium |
3.75 DKK
Pack of 100 |
In Stock
Log in to see stock. |
|||||
16806081
|
AMPri™
01037L |
Large |
15.00 DKK
Pack of 100 |
Estimated Shipment: 27-11-2025
Log in to see stock. |
|||||
16836081
|
AMPri™
01037XL |
X-Large |
4.75 DKK
Pack of 100 |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
Description
- AQL 1.5
- Premium grip examination gloves
- Applications include laboratory, clinical, pharmaceutical, dental, and food industry use
- Protect against chemicals and microorganisms
- Ambidextrous fit
- Powder-free
- Non-sterile
- Color: white
- Textured finish
- Disposable
Specifications
AQL 1.5 | |
White | |
Laboratory | |
0.08 mm (Cuff) |
EN 455-1, EN374-1/4/5, EN420, Food grade | |
Single-use | |
No | |
240 mm |