Learn More
Mercury, Electronic Grade, 99.9998% (metals basis)
Product Code. p-7048351
Click to view available options
Quantity:
2270 g
4540 g
Unit Size:
2270g
4540g
CAS: 7439-97-6 | Hg | 200.59 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7439-97-6
Molecular Formula Hg
Molecular Weight (g/mol) 200.59
MDL Number MFCD00011035
InChI Key QSHDDOUJBYECFT-UHFFFAOYSA-N
PubChem CID 23931
ChEBI CHEBI:16170
IUPAC Name mercury
SMILES [Hg]
Specifications
Color Silver
Assay Percent Range 99.9998% (Metals basis)
UN Number UN2809
Quantity 2270 g
Synonym hydrargyrum, quicksilver, metallic, liquid silver, mercurio, quecksilber, mercure, quick silver, colloidal, mercury, metallic
Percent Purity 99.9998%
Grade Electronic
Odor Odorless
Form Liquid
Assay (metals basis)
Chemical Name or Material Mercury