Learn More
Cole-Parmer™ Mesh Shelf
Static stainless steel Mesh Shelf for use in Techne hybridizers, able to support up to 4 kg in weight.
Brand: Cole-Parmer™ 39455-26
1483.24 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Can be placed into the tube locators.
- Allows the unit to be used as a simple non-motion incubator.
- For use with either HB-1D or HB-3D hybridizers.
Specifications
HB-1D Hybridizer | |
Up to 4 kg | |
Stainless Steel | |
Shelf |