Learn More
Methanol, 99.9%, Anhydrous, Scharlab
Product Code. 12406232
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle with Cap/Septum
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High purity solvent with minimal water content (max. 0.003% H2O). Methanol, 99.9%, Anhydrous, Scharlab is a ready to use, high-purity solvent available without a molecular sieve.
Anhydrous solvents are essential in organic, organometallic and oligonucleotide synthesis and in inorganic chemistry, where many reactions must take place in moisture-free conditions. Anhydrous solvents are also frequently used in combinatorial chemistry and in biotechnology.
  • Avoids the hazards of preparing anhydrous solvents via distillation over metals
  • Guaranteed quality by means of strict Karl Fischer controls
Specifications
Melting Point -98°C
CAS 67-56-1
Color Colorless
Acidity Max. 0.0002meq/g
Alkalinity Max. 0.0002meq/g
Appearance Clear
Density 0.79g/cm3
Boiling Point 64.5°C
Quantity 1 L
Flash Point 11°C
Identification Passes test
Ignition Point 455°C
Packaging Glass Bottle with Cap/Septum
Physical Form Liquid
Refractive Index 1.3288
Residue after Evaporation Max. 0.0005%
Molecular Formula CH4O
Linear Formula CH3OH
MDL Number MFCD00004595
Vapor Pressure 128 hPa
UN Number 1230
Water Max. 0.003%
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
Solubility Information Miscible in water
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
PubChem CID 887
ChEBI CHEBI:17790
Formula Weight 32.04g/mol
Percent Purity Min. 99.9%
Grade Anhydrous
Chemical Name or Material Methanol