Learn More
Methanol HPLC Grade, Carl Roth™
Click to view available options
Quantity:
1 L
Packaging:
Glass
Specifications
Specifications
Density | 0.79 |
Boiling Point | 64.7°C |
Quantity | 1 L |
Packaging | Glass |
Concentration or Composition (by Analyte or Components) | 1L |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.