Learn More
Methylene Blue, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
1105.00 DKK
Chemical Identifiers
CAS | 7220-79-3 |
---|---|
Molecular Formula | C16H24ClN3O3S |
Molecular Weight (g/mol) | 373.90 |
MDL Number | MFCD00150008 |
InChI Key | XQAXGZLFSSPBMK-UHFFFAOYSA-M |
Synonym | C.I. 52015 |
PubChem CID | 104827 |
SMILES | O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15578814
|
Avantor J.T.Baker
Q473-03 |
25 g | Wide Mouth Amber Glass Bottle |
1105.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7220-79-3 | |
373.90 | |
XQAXGZLFSSPBMK-UHFFFAOYSA-M | |
104827 |
C16H24ClN3O3S | |
MFCD00150008 | |
C.I. 52015 | |
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C |
Specifications
7220-79-3 | |
MFCD00150008 | |
XQAXGZLFSSPBMK-UHFFFAOYSA-M | |
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;trihydrate | |
104827 | |
BAKER ANALYZED™ Reagent |
C16H24ClN3O3S | |
C.I. 52015 | |
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C | |
373.90 | |
373.91 | |
Methylene Blue |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Methylene blue
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Methylene blue