Learn More
Miele Professional™ Miele Process Documentation
APH 302 Connection cable type 2 for connection of a machine with serial interface for a PC/Net500.
Brand:  Miele Professional™ 7951410
232.52 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15470445
  • 324.00 DKK / Each
Only null left
Estimated Shipment: 29-09-2025
Description
Compatible with appliances from Sirona and W and H (cable length 3m)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Process/Operating Documentation/Communication
Washer-disinfectors, laboratory