Learn More
Miele Professional™ Miele Process Documentation
APH 406 Network cable for connection to the network, length 3 m (type 6).
Brand: Miele Professional™ 7951470
232.52 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Cable category 6 (color: transparent silver)
Specifications
Process/Operating Documentation/Communication | |
Washer-disinfectors, laboratory |