Learn More
Brady™ MiniMark™ Labeling Ribbons
These ribbons have standard scratch, smear and chemical resistance. MiniMark™ Labeling Ribbons for B-7951 and B-7589 materials.
Brand: Brady™ 710024
This item is not returnable.
View return policy
Description
- These ribbons have standard scratch, smear and chemical resistance MiniMark ribbon for B-7951 and B-7589 materials.
- MiniMark ribbon for B-7951 and B-7589 materials.
Specifications
Printer Ribbon | |
MiniMark Printer | |
4.3 in. | |
110 mm | |
12.7 mm I.D. | |
110 mm Red Ribbon (2 Pk.) | |
General purpose ribbon for all manners of Labeling |
Red | |
98.4 yd. | |
90 m | |
0.5 in. ID | |
90 m x 110 mm | |
Ink side out | |
2 Pk. |