Learn More
Sigma Laborzentrifugen™ Multiple Carrier for Centrifuge Tubes
Holds a variety of tube types securely during the centrifuging process.
212.00€ - 1780.00€
Product Code | Brand | Capacity | For Use With (Equipment) | Tube Type | Max. Tube Length | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | For Use With (Equipment) | Tube Type | Max. Tube Length | Price | Quantity & Availability | ||
11700739
|
Sigma Laborzentrifugen™
13033 |
5 x 5 mL Tubes | 11030, Swing-out rotor | RIA, Vacutainer, Hemolyse Tubes, Round and Flat Bottom | 65 to 85 mm | N/A | |||
10297043
|
Sigma Laborzentrifugen™
13037 |
6 x 5 to 15 mL Tubes | 11030, Swing-out rotor | Round Bottom, Monovettes | 90 to 105 mm |
212.00€
Pack of 2 |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||
11790729
|
Sigma Laborzentrifugen™
13233 |
50 x 5 mL Tubes | 11150, Swing-out rotor, 11151, Swing Out Rotor 48 x 15mL complete, no 11150 incl 2 sets rectangular buckets no 13215, 2 sets rectangular carriers no 18015 and 48 PS-tubes 15mL no 15020 | RIA | 70 to 90 mm | N/A | |||
10209441
|
Sigma Laborzentrifugen™
13235 |
7 x 50 mL Tubes | 11150, Swing-out rotor, 11151, Swing Out Rotor 48 x 15mL complete, no 11150 incl 2 sets rectangular buckets no 13215, 2 sets rectangular carriers no 18015 and 48 PS-tubes 15mL no 15020 | Culture |
1240.00€
Pack of 2 |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||
10148011
|
Sigma Laborzentrifugen™
13009 |
5 x 5 mL Tubes | 11390, Swing-out rotor for 4 buckets, 11190, Swing-out rotor 4 x 100mL complete, 11191, Swing-out rotor 16 x 15mL complete, 11192, Swing-out rotor for 4 | RIA, Vacutainer, Hemolyse Tubes, Round and Flat Bottom | 65 to 85 mm |
346.00€
Pack of 2 |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||
Description
- Holds a variety of tube sizes and capacities
- Provided in a set of 2
Specifications
Adapter | |
2/Pk. |