Learn More
Macherey-Nagel™ N20 Aluminum Closures
Shop All Macherey-Nagel Products
Click to view available options
Color:
Gold
Silver
Septum:
Butyl
Butyl/PTFE
Silicone/PTFE
Color (Seal):
Dark Gray
Dark Gray/Gray
Gray
White/Beige
Type:
Center Tear-off Crimp Cap
Complete Tear-off Crimp Cap
Crimp Cap
Pressure Release Crimp Cap
Unit Size:
Each
Pack of 100
Description
- Center tear off caps and complete tear off caps N 20
- Bi-metal crimp caps and magnetic crimp caps N 20
- for use on autosamplers with magnetic transportation system –
- Crimping of the bi-metal crimp caps is more convenient.
- All closures mentioned above are available with silicone / PTFE septa as well as with butyl / PTFE septa in different versions.
- Applications above 120 °C require silicone / PTFE septa.
- Individual septa, stoppers as well as PE caps N 20 are also available.
- Manual and electronic crimping tools N 20 complete this product range
Specifications
Specifications
Color (Seal) | Dark Gray/Gray |
Color | Gold |
Material | Aluminum, Butyl Rubber, PTFE |
Septum | Butyl/PTFE |
Quantity | 100 Pk. |
Thickness (Metric) | 3 mm |
Type | Center Tear-off Crimp Cap |