Learn More
Rogo Sampaic™ Nageotte Counting Cells
For determining the number of particles per volume unit of a liquid. Rogo Sampaic™ Nageotte Counting Cells have a simple or double grid with a depth of 0.5mm.
177.29 € - 182.69 €
Specifications
Product Type | Nageotte Cell |
---|---|
Quantity | 1Each |
Sample Type | Spinal, Urinary |
Depth (Metric) | 0.5mm |
Description
- For spinal and urinary counts
- Provides for accurate manual measurements
Specifications
Nageotte Cell | |
1Each | |
Spinal, Urinary | |
0.5mm |